Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Mouse IgG3 Secondary Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB554262 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB554262 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB554262 Supplier Novus Biologicals Supplier No. NBP268520PCP
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG3 Polyclonal specifically detects IgG3 in Mouse samples. It is validated for ELISA.

Specifications

Antigen IgG3
Applications ELISA
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809
Host Species Goat
Immunogen Mouse IgG3 paraproteins
Purification Method Affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3502
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.