Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Goat anti-Mouse IgM, H + L Chain Specific, Peroxidase, Secondary Antibody,

Catalog No. 4012252ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mL
Conjugate:
Peroxidase
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
40-122-52ML 2 mL Peroxidase
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-122-52ML Supplier MilliporeSigma™ Supplier No. 4012252ML

Goat Polyclonal Antibody

Specifically detects IgM, H&L Chain Specific Clone: in Mouse samples, and it is validated for Enzyme Immunoassay, Immunoelectrophoresis

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgM, H&L Chain Specific
Applications ELISA, Immunoelectrophoresis
Classification Polyclonal
Conjugate Peroxidase
Formulation In 10mM PBS, 0.5mM EDTA, 10% mannose, 2% sucrose, 1% BSA, pH 7.6.
Host Species Goat
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2°C to 8°C
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.