Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Mouse IgM Heavy Chain Secondary Antibody, DyLight 800, Novus Biologicals™
SDP

Catalog No. NBP172884 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP172884 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP172884 Supplier Novus Biologicals Supplier No. NBP172884
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgM Heavy Chain Polyclonal specifically detects IgM Heavy Chain in Mouse samples. It is validated for Fluorophore-linked immunosorbent assay, Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen IgM Heavy Chain
Applications Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate DyLight 800
Dilution Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:5000, Fluorophore-linked immunosorbent assay 1:20000
Gene Alias Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain
Gene Symbols IGHM
Host Species Goat
Immunogen Mouse IgM u chain
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3507
Test Specificity No reaction was observed against other mouse heavy or light chain proteins.
Reconstitution Reconstitute with 100 ul deionized water (or equivalent).
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.