Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Mouse IgM Heavy Chain Secondary Antibody, HRP (Pre-adsorbed), R&D Systems™
SDP

Catalog No. NBP175193 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP175193 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP175193 Supplier R&D Systems Supplier No. NBP175193
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Goat Polyclonal Antibody

IgM Heavy Chain Polyclonal specifically detects IgM Heavy Chain in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen IgM Heavy Chain
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate HRP
Dilution Western Blot 1:200-1:5000, ELISA 1:200-1:5000, Immunohistochemistry 1:500-1:5000, Immunohistochemistry-Paraffin
Gene Alias Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain
Host Species Goat
Immunogen Purified Mouse IgM, u chain
Purification Method Affinity purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3507
Test Specificity Based on IEP, this antibody reacts with heavy mu chains on mouse IgM. This antibody has been pre-adsorbed against human IgG and serum proteins.
Reconstitution Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily.
Target Species Mouse
Content And Storage Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage.
Product Type Antibody
Isotype IgM
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.