Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat F(ab')2 Anti-mouse IgM-PerCP Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. F0119 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0119 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. F0119 Supplier Novus Biologicals Supplier No. F0119
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

IgM Polyclonal specifically detects IgM in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen IgM
Applications Flow Cytometry
Classification Polyclonal
Conjugate PerCP
Dilution Flow Cytometry
Gene Alias AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH
Gene Symbols IGHM
Host Species Goat
Immunogen Mouse IgM heavy and light chains
Purification Method Antigen Affinity-purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3507
Test Specificity The goat IgG is first purified by affinity chromatography and then adsorbed to eliminate human cross reactivity. The IgG fraction is digested with pepsin to generate F(ab')2 fragments which have a reduced ability to interact with Fc receptors expressed on a variety of cells.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date. Protect from light.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.