Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Mouse Kappa Light Chain Secondary Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB7548 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB7548 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB7548 Supplier Novus Biologicals Supplier No. NB7548
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Kappa Light Chain Polyclonal specifically detects Kappa Light Chain in Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen Kappa Light Chain
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate FITC
Dilution Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunohistochemistry-Paraffin
Gene Alias HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809
Gene Symbols IGKC
Host Species Goat
Immunogen Mouse Kappa Light Chain
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3514
Test Specificity By immunoelectrophoresis and ELISA this reacts specifically with kappa light chains common to all mouse immunoglobulins. No was detected against lambda light chain or non-immunoglobulin serum proteins.
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.