Learn More
Goat anti-Mouse Kappa Light Chain Secondary Antibody, HRP, R&D Systems™
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Antigen | Kappa Light Chain |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | HRP |
| Dilution | Western Blot 1:2000 - 1:20000, ELISA 1:10000-1:100000, Immunohistochemistry 1:200 - 1:2000, Immunocytochemistry/Immunofluorescence 1:200-1:500, Immunohistochemistry-Paraffin 1:200 - 1:2000 |
| Gene Alias | HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 |
| Gene Symbols | IGKC |
| Host Species | Goat |
| Immunogen | Mouse Kappa Light Chain |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.