Learn More
Goat anti-Mouse Lambda Light Chain Secondary Antibody, DyLight 755, R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | Lambda Light Chain |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 755 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | IGLC, immunoglobulin lambda constant group |
| Host Species | Goat |
| Immunogen | Mouse Lambda Light Chain |
| Purification Method | Immunogen affinity purified |
| Quantity | 1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.