Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Porcine IgA Heavy Chain Secondary Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB727 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB727 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB727 Supplier Novus Biologicals Supplier No. NB727
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

IgA Heavy Chain Polyclonal specifically detects IgA Heavy Chain in Pig samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen IgA Heavy Chain
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate HRP
Dilution Western Blot 1:2000 - 1:20000, ELISA 1:10000- 1:100000, Immunohistochemistry 1:200 - 1:2000, Immunocytochemistry/Immunofluorescence 1:200 - 1:2000, Immunohistochemistry-Paraffin 1:200 - 1:2000
Gene Alias Immunoglobulin A
Gene Symbols IGHA1
Host Species Goat
Immunogen Pig IgA
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3493
Test Specificity By immunoelectrophoresis and ELISA this reacts specifically with pig IgA. This may cross react with IgA from other species.
Target Species Pig
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.