Learn More
Goat anti-Porcine IgG Fc Secondary Antibody, Allophycocyanin, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | IgG Fc |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | APC |
| Formulation | PBS |
| Host Species | Goat |
| Immunogen | Pig IgG-Fc Fragment |
| Purification Method | Immunogen affinity purified |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.