Learn More
Goat anti-Rabbit IgA Heavy Chain Secondary Antibody, DyLight 680, R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgA Heavy Chain |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Host Species | Goat |
| Immunogen | Rabbit IgA |
| Purification Method | Immunogen affinity purified |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.