Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Goat anti-Rabbit, Brilliant Violet 421, Polyclonal, BD

Catalog No. BDB565014 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565014 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565014 Supplier BD Biosciences Supplier No. 565014
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

BD Horizon™ BV421 Goat Anti-Rabbit IgG is intended to be a second-step reagent for immunofluorescent staining of cells pre-stained with Rabbit IgG primary antibodies. It reacts with rabbit IgG and the light chains of other rabbit immunoglobulins. BV421 Anti-Rabbit IgG has minimal crossreactivity with human, mouse, or rat serum proteins. As a second step staining reagent, it is reactive with rabbit polyclonal and monoclonal antibodies.

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. BD Horizon BV421 has an Ex Max at 407nm and Em Max at 421 nm. The use of a mounting reagent (eg, ProLong™ Gold) is highly recommended to maximize the photostability of BV421.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications Flow Cytometry, Immunofluorescence
Classification Polyclonal
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Goat
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype Ig
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.