Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Rabbit IgG F(ab')2 Secondary Antibody, R&D Systems™
SDP

Catalog No. NBP26065910 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP26065910 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP26065910 Supplier R&D Systems Supplier No. NBP26065910MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Goat Polyclonal Antibody

IgG Polyclonal specifically detects IgG in Rabbit samples. It is validated for ELISA, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:1000-1:5000, Fluorophore-linked immunosorbent assay 1:10000-1:50000
Formulation 0.01M Sodium Phosphate, 0.15M Sodium Chloride, pH 7.2 with 10Mg/ml BSA - Immunoglobulin and Protease free with No Preservative
Gene Alias immunoglobulin heavy constant gamma 1 (G1m marker)
Gene Symbols IGHG
Host Species Goat
Immunogen Rabbit IgG F(ab')2 fragment
Purification Method Ion exchange chromatography
Quantity 10 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity This product is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Goat Serum, Rabbit IgG, Rabbit IgG F(ab')2 and Rabbit Serum. No reaction was observed against Rabbit IgG F(c).
Reconstitution Reconstitute with 0.1mL deionized water.
Target Species Rabbit
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.