Learn More
Description
Specifications
Specifications
| Antigen | IgG Fc |
| Applications | ELISA, Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Formulation | PBS |
| Host Species | Goat |
| Immunogen | Goat anti-Rabbit IgG Fc fragment Secondary Antibody was produced by repeated immunization with rabbit IgG f(c) fragment in goat |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.5 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
