Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Rabbit IgG Fc Secondary Antibody, Novus Biologicals™
SDP

Catalog No. NBP175342 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP175342 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP175342 Supplier Novus Biologicals Supplier No. NBP175342
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG Fc Polyclonal specifically detects IgG Fc in Rabbit samples. It is validated for ELISA, Immunoassay, Immunodiffusion, Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen IgG Fc
Applications ELISA, Immunoassay, Immunodiffusion, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 4.5mg/mL
Conjugate Unconjugated
Dilution Immunoassay
Host Species Goat
Immunogen Purified Rabbit IgG, Fc fragment
Purification Method Affinity purified
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity Based on IEP, this antibody reacts with heavy gamma chains on rabbit IgG
Target Species Rabbit
Content And Storage Store at 4C. After dilution, add an equal volume of glycerol to prevent loss of enzymatic activity and store at -20C. Avoid repeated freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.