Learn More
MilliporeSigma™ Goat anti-Rabbit IgG (H,L), Alexa Fluor 488, Polyclonal Secondary Antibody,
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Antigen | Goat anti-Rabbit IgG (HL) |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1.5mg |
| Conjugate | Alexa Fluor 488 |
| Formulation | Lyophilized in buffer containing 0.01M Sodium Phosphate, 0.25M NaCl, pH7.6 with 15mg/mL BSA (IgG-Free, Protease-Free) and 0.05% sodium azide. |
| Host Species | Goat |
| Purification Method | Affinity Purified |
| Quantity | 1.5 mg |
| Primary or Secondary | Secondary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.