Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Rabbit IgG (H+L) Secondary Antibody, Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB010670 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010670 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010670 Supplier Novus Biologicals Supplier No. NBP279817APCCY7
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay.

Specifications

Antigen IgG (H+L)
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay
Classification Polyclonal
Conjugate APC-Cyanine7
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay
Formulation PBS with 0.05% Sodium Azide
Host Species Goat
Immunogen Purified Rabbit IgG (H&L)
Purification Method Affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity Based on Immunoelectrophoresis, this antibody reacts with heavy (gamma) chains on rabbit IgG and light chains on all rabbit immunoglobulins. Cross Adsorbed: This antibody was pre-adsorbed to reduce cross-reactivity with proteins of non-related species. Minimal cross-reactivity with bovine, human, mouse IgG serum proteins.
Target Species Rabbit
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.