Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Rabbit IgG (H+L) Secondary Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP172957 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP172957 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP172957 Supplier Novus Biologicals Supplier No. NBP172957
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Fluorophore-linked immunosorbent assay, Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen IgG (H+L)
Applications Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate DyLight 405
Dilution Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:5000, Fluorophore-linked immunosorbent assay 1:20000
Host Species Goat
Immunogen Rabbit IgG whole molecule
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity This antibody will react with heavy chains of Rabbit IgG and with light chains of most Rabbit immunoglobulins.
Reconstitution Reconstitute with 100 ul deionized water (or equivalent).
Target Species Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.