Learn More
Goat anti-Rabbit IgG (H+L) Secondary Antibody, DyLight 680, R&D Systems™
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 680 |
| Formulation | 50mM Sodium Borate |
| Host Species | Goat |
| Immunogen | This Goat anti-Rabbit IgG (H+L) Secondary Antibody (Pre-adsorbed) was developed against rabbit IgG. |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.5 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.