Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Rabbit IgG (H+L) Secondary Antibody, HRP (Pre-adsorbed), Novus Biologicals™
SDP

Catalog No. NBP17530401 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17530401 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17530401 Supplier Novus Biologicals Supplier No. NBP1753040.1MG
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Dot Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunomicroscopy, Western Blot.

Specifications

Antigen IgG (H+L)
Applications Dot Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunomicroscopy, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate HRP
Dilution Western Blot 1:200-1:5000, ELISA 1:200-1:5000, Immunohistochemistry 1:500-1:5000, Immunohistochemistry-Paraffin, Immunomicroscopy
Formulation 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free. with 0.1% Kathon
Host Species Goat
Immunogen Purified Rabbit IgG (H&L)
Purification Method Affinity Purified
Quantity 0.1 mg
Primary or Secondary Secondary
Test Specificity Based on Immunoelectrophoresis, this antibody reacts with heavy (gamma) chains on rabbit IgG and light chains on all rabbit immunoglobulins. Cross Adsorbed: This antibody was pre-adsorbed to reduce cross-reactivity with proteins of non-related species. Minimal cross-reactivity with bovine, human, mouse IgG serum proteins.
Reconstitution Reconstitute with 1.1 ml deionized water and let stand 30 minutes at RT to disolve. Centrifuge to remove any particulates. The 0.1 mg size will be shipped in liquid.
Target Species Rabbit
Content And Storage Store at 4C. After reconstitution, dilute with 50% glycerol and store at -20C. Avoid repeated freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.