Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Rabbit IgG (Heavy Chain), Superclonal™ Recombinant Secondary Antibody, Alexa Fluor™ 680, Invitrogen™

Catalog No. PIA27042
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA27042 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIA27042 Supplier Thermo Scientific Supplier No. A27042
Add to Cart
Add to Cart

Goat Recombinant Polyclonal Secondary Antibody

The sensitivity and specificity of each lot is confirmed using Western blot. Minimal cross-reactivity with rabbit, mouse, rat, human, bovine, guinea pig and donkey IgG is observed. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency.

Rabbit IgG (Heavy Chain) Recombinant Polyclonal antibody specifically detects Rabbit IgG (Heavy Chain) in Rabbit samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabbit IgG (Heavy Chain)
Applications Western Blot
Classification Recombinant Polyclonal
Concentration 1 mg/mL
Conjugate Alexa Fluor 680
Formulation PBS with 5mM sodium azide; pH 7.2
Host Species Goat
Immunogen Recombinant full-length protein
Purification Method purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 4° C, store in dark, DO NOT FREEZE!
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.