Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Goat anti-Rabbit IgG , Secondary Antibody ™ (Chemicon™)
Supplier: MilliporeSigma™ AP132B/DEL
Description
Isotype: IgG
Host Species: Goat
Cross Reactivity: Rabbit
Format: Purified
ELISA, Immunoelectrophoresis, Immunoprecipitation

Specifications
Goat anti-Rabbit IgG | |
Biotin | |
Affinity Purified | |
RUO | |
Secondary |
Polyclonal | |
Goat | |
2 mL | |
Enzyme | |
Rabbit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction