Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat Anti-Rat F(ab)2 IgG (H+L) PerCP-conjugated Antibody, R&D Systems™
Goat Polyclonal Antibody
Supplier: R&D Systems F0115
This item is not returnable.
View return policy
Description
IgG Polyclonal specifically detects IgG in Rat samples. It is validated for Flow Cytometry.Specifications
IgG | |
Polyclonal | |
Flow Cytometry | |
IGHG1 | |
Rat IgG heavy and light chains | |
100 Tests | |
Secondary | |
Detects rat IgG primary antibodies in immunofluorescent assays. After purification by affinity chromatography, this antibody was adsorbed to eliminate mouse cross-reactivity and the IgG fraction was further digested with pepsin to generate F(ab') 2 fragments which have reduced interactions with cell surface Fc receptors. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Flow Cytometry | |
PerCP | |
Immunoglobulin G, ImmunoglobulinG | |
Goat | |
Affinity Purified | |
RUO | |
3500 | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction