Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat anti-Rat IgA/IgG/IgM, H/L Chains Antibody, AP, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP172661
Description
IgA/IgG/IgM (H+L) Polyclonal specifically detects IgA/IgG/IgM (H+L) in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
IgA/IgG/IgM (H+L) | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Goat | |
Affinity Purified | |
RUO | |
This reagent is suitable for the detection of all Rat immunoglobulin isotypes, subclasses and chain combinations. | |
Store at 4C. Do not freeze. |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1:1000 - 1:5000, ELISA 1:2000 - 1:10000, Immunohistochemistry 1:500 - 1:2500 | |
Rat IgG, IgA and IgM whole molecule | |
1 mg | |
Secondary | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction