Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat anti-Rat IgA/IgG/IgM, H/L Chains Antibody, Fluorescein, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP173486
Description
IgA/IgG/IgM (H+L) Polyclonal specifically detects IgA/IgG/IgM (H+L) in Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.Specifications
IgA/IgG/IgM (H+L) | |
Polyclonal | |
Fluorescein | |
Goat | |
Affinity Purified | |
RUO | |
This reagent is suitable for the detection of all Rat isotypes and chain combinations. | |
Rat | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
LYOPH | |
Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
Rat IgG, IgA and IgM whole molecules | |
2 mg | |
Secondary | |
Reconstitute with 1.0 ml deionized water (or equivalent). | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction