Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Goat anti-Rat IgG, Alexa Fluor 647, Polyclonal Secondary Antibody,

Catalog No. AP183SA6MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP183SA6MI 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP183SA6MI Supplier MilliporeSigma™ Supplier No. AP183SA6

Specifications

Antigen Goat anti-Rat IgG
Classification Polyclonal
Concentration 0.5mg
Conjugate Alexa Fluor 647
Formulation Lyophilized in buffer containing 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA (IgG-Free, Protease-Free) and 0.05% sodium azide.
Application: Reconstitute the product to 0.4mLwith dH2O. Suggested dilution range for most applications: 1:1
Host Species Goat
Purification Method Affinity Purified
Quantity 0.5 mg
Primary or Secondary Secondary
Target Species Rat
Content And Storage 2-8°C for up to 12 months.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.