Learn More
Goat anti-Rat IgG F(Ab')2 Polyclonal Secondary Antibody, MP Biomedicals
Shop All MP Biomedicals, Inc ProductsDescription
Host Species: Goat
Isotype: IgG
Species Reactivity: Rat
Specifications
Specifications
| Antigen | IgG F(Ab')2 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Goat |
| Quantity | 5 mL |
| Primary or Secondary | Secondary |
| Target Species | Rat |
| Form | Purified |
| Isotype | IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.