Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat Anti-Rat IgG NorthernLights™ NL493-conjugated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. NL015 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NL015 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NL015 Supplier R&D Systems Supplier No. NL015
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

IgG Polyclonal specifically detects IgG in Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry.

Specifications

Antigen IgG
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration 1mg/mL
Conjugate NorthernLights 493 (NL493)
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Goat
Purification Method Affinity Purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Target Species Rat
Content And Storage Store the unopened product at 2 - 8degrees C. Do not use past expiration date. Protect from light.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.