Learn More
Enzo Life Sciences Goat anti-rat IgG, polyclonal antibody (HRP conjugate) (0.5 ml)
SDP
Supplier:  Enzo Life Sciences ADISAB2200500
Encompass_Preferred
Host: Goat. Immunogen: Purified rat IgG (whole molecule). Species Reactivity: Rat. Specificity: Based on immunoelectrophoresis and/or ELISA, the antibody reacts with whole molecule rat IgG. It also reacts with the light chains of other rat immunoglobulins. No antibody was detected against non-immunoglobulin serum proteins. Applications: ELISA, ICC, IHC, WB. Recommended dilutions: ELISA (1:5,000-1:100,000). Immunocytochemistry (1:500-1:5,000). Immunohistochemistry(1:500-1:5,000). Western Blot (1:10,000-1:200,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Immunoaffinity purified. Formulation: Liquid. In 0.005M sodium phosphate, pH 7.6, containing 0.125M sodium chloride, 7.5mg/ml BSA (IgG-free, protease-free), and 50% glycerol. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-103-1713
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.