Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Rat IgG, Whole Molecule Polyclonal Secondary Antibody, HRP, MP Biomedicals

$725.00

Specifications

Antigen IgG, Whole Molecule
Classification Polyclonal
Conjugate HRP
Host Species Goat
Target Species Rat
View More Specs

Products 1
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
ICN55741
View Documents
MP Biomedicals, Inc
0855741
Purified 2 mg
Each of 1 for $725.00
Only null left
Add to Cart
 
Description

Description

Host Species: Goat
Species Reactivity: Rat

Specifications

Specifications

IgG, Whole Molecule
HRP
Rat
Polyclonal
Goat
Secondary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.