Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Rat IgG1 Heavy Chain Secondary Antibody, FITC, R&D Systems™
SDP

Catalog No. NB7119 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB7119 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB7119 Supplier R&D Systems Supplier No. NB7119
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Goat Polyclonal Antibody

IgG1 Heavy Chain Polyclonal specifically detects IgG1 Heavy Chain in Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen IgG1 Heavy Chain
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate FITC
Dilution Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunohistochemistry-Paraffin 1:50 - 1:500
Host Species Goat
Immunogen Rat IgG1
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity By immunoelectrophoresis and ELISA this reacts specifically with Rat IgG1. This may cross react with IgG1 from other species.
Target Species Rat
Content And Storage Store at 4C. Do not freeze.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.