Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Rat IgG1 Secondary Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB551993 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB551993 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB551993 Supplier Novus Biologicals Supplier No. NBP268483R
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG1 Polyclonal specifically detects IgG1 in Rat samples. It is validated for ELISA, Flow Cytometry, Western Blot.

Specifications

Antigen IgG1
Applications ELISA, Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias IgG1, Igh-4, RGD1359539, VH7183
Host Species Goat
Immunogen Rat IgG1
Purification Method Affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.