Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Rat IgG1 Secondary Antibody, HRP, R&D Systems™
SDP

Catalog No. NB002841 Shop All Novus Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002841 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002841 Supplier R&D Systems Supplier No. NBP268485500UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Goat Polyclonal Antibody

IgG1 Polyclonal specifically detects IgG1 in Rat samples. It is validated for ELISA, Western Blot.

Specifications

Antigen IgG1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Dilution Western Blot 1:2000 - 1:20000, ELISA 1:10000 - 1:100000
Formulation PBS, 0.2% BSA
Host Species Goat
Immunogen Rat IgG1
Purification Method Affinity Purified
Quantity 500 μg
Regulatory Status RUO
Test Specificity Goat anti Rat IgG1 has been shown to react with rat IgG1 by immunoelectrophoresis and ELISA. The antibody may cross-react with IgG1 from other species.
Target Species Rat
Content And Storage Store at 4°C.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.