Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Rat IgG1 Secondary Antibody, PE, R&D Systems™
SDP

Catalog No. NB551282 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB551282 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB551282 Supplier R&D Systems Supplier No. NBP268483PE
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Goat Polyclonal Antibody

IgG1 Polyclonal specifically detects IgG1 in Rat samples. It is validated for ELISA, Flow Cytometry, Western Blot.

Specifications

Antigen IgG1
Applications ELISA, Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias IgG1, Igh-4, RGD1359539, VH7183
Host Species Goat
Immunogen Rat IgG1
Purification Method Affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.