Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat anti-Rat IgG2a Heavy Chain Secondary Antibody, Alexa Fluor™ 488, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB7122AF488
Description
IgG2a Heavy Chain Polyclonal antibody specifically detects IgG2a Heavy Chain in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
IgG2a Heavy Chain | |
Polyclonal | |
Alexa Fluor 488 | |
Goat | |
Immunogen affinity purified | |
RUO | |
Secondary | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
50mM Sodium Borate | |
Rat IgG2a | |
1 mL | |
Adaptive Immunity, Immunology | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction