Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Rat IgG2a Secondary Antibody [Janelia Fluor 669], Novus Biologicals™
SDP

Catalog No. NB786116 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB786116 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB786116 Supplier Novus Biologicals Supplier No. NBP268486JF669
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG2a Polyclonal antibody specifically detects IgG2a in Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA

Specifications

Antigen IgG2a
Applications Western Blot, Flow Cytometry, ELISA
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Host Species Goat
Immunogen Rat IgG2a
Purification Method Affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.