Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Rat IgG2b Secondary Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB002843 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002843 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002843 Supplier Novus Biologicals Supplier No. NBP268491500UG
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG2b Polyclonal specifically detects IgG2b in Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen IgG2b
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Dilution Western Blot 1:1000 - 1:30000, ELISA 1:10000 - 1:100000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500, Immunohistochemistry-Frozen 1:200 - 1:500
Formulation PBS, 0.2% BSA
Host Species Goat
Immunogen Rat IgG2b
Purification Method Affinity Purified
Quantity 500 μg
Test Specificity Goat anti Rat IgG2b antibody recognizes rat IgG2b as assessed by immunoelectrophoresis and ELISA. The antibody may cross-react with IgG2b from other species.
Target Species Rat
Content And Storage Store at 4°C.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.