Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDFY1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132537 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-537 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-537 Supplier Abnova Corporation Supplier No. PAB6612
Only null left
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of WDFY1.

The FYVE domain mediates the recruitment of proteins involved in membrane trafficking and cell signaling to phosphatidylinositol 3-phosphate (PtdIns(3)P)-containing membranes. This gene encodes a protein which contains a single FYVE domain and multiple WD40 repeats. This protein is localized to endosomes. [provided by RefSeq

Sequence: VVGCSLATGFSPH

Specifications

Antigen WDFY1
Applications ELISA, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of WDFY1.
Dilution ELISA (1:32000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-6 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene WDFY1
Gene Alias FENS-1/WDF1/ZFYVE17
Gene Symbols WDFY1
Host Species Goat
Immunogen A synthetic peptide corresponding to C-terminus of human WDFY1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57590
Test Specificity This antibody is expected to recognize both reported isoforms (as represented by NP_065881 and NP_848127)
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.