Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Goat BMP-4 ELISA Kit (Chemiluminescence)
SDP

Catalog No. NBP342686 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP342686 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP342686 Supplier Novus Biologicals™ Supplier No. NBP342686
Add to Cart
Add to Cart

Chemiluminescence ELISA Kit

Specifications

Assay Range 27.4 to 20000 pg/mL
Assay Sensitivity 12.3 pg/mL
Detection Method Colorimetric
Immunoassay Kit Format Pre-coated 96T Strip Plate
Sample Type Serum, Plasma, Tissue Homogenates, Other Biological Fluids
Target Species Goat
For Use With (Application) ELISA
Gene ID (Entrez) 652
Gene Symbol BMP4
Kit Contents Pre-coated 96T Strip Plate, Plate sealer for 96 wells, Standard, Standard Diluent, Detection Reagent A, Assay Diluent A, Detection Reagent B, Assay Diluent B, Substrate A, Substrate B, Wash Buffer (30X concentrate), Instruction manual
Quantity 1 Kit
Research Discipline Biologically Active Proteins, Stem Cell Markers, Stem Cell Signaling Pathway
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym BMP-2B, BMP2B1, BMP2BMCOPS6, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, OFC11, ZYME
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.