Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat F(ab')2 anti-Mouse Complement C3 Polyclonal Anitbody, MP Biomedicals
Supplier: MP Biomedicals, Inc 0855474
Description
Host Species: Goat
Species Reactivity: Mouse

Specifications
Complement C3 | |
Unconjugated | |
2 mL | |
Mouse |
Polyclonal | |
Goat | |
Primary | |
Lyophilized |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction