Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat F(ab')2 anti-Mouse IgG Fc Secondary Antibody, FITC (Pre-adsorbed), R&D Systems™
SDP

Catalog No. NB7107817 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB7107817 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB7107817 Supplier R&D Systems Supplier No. NB7107817
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Goat Polyclonal Antibody

IgG Fc Polyclonal specifically detects IgG Fc in Mouse samples. It is validated for Dot Blot, Flow Cytometry, Fluorophore-linked immunosorbent assay, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen IgG Fc
Applications Dot Blot, Flow Cytometry, Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate FITC
Dilution Flow Cytometry 1:500 - 1:2500, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Dot Blot, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000
Host Species Goat
Immunogen Mouse IgG F(c) fragment
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity No reaction was observed against anti-Pepsin, anti-Goat IgG F(c), Mouse IgG F(ab) or Bovine, Horse and Human Serum Proteins.
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.