Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat F(ab')2 anti-Mouse IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™

Goat Polyclonal Antibody
$366.00
Specifications
Antigen | IgG (H+L) |
---|---|
Concentration | 4.5mg/mL |
Dilution | Flow Cytometry 1:20-1:2000, Immunomicroscopy 1:20-1:2000 |
Applications | Flow Cytometry, Immunomicroscopy |
Classification | Polyclonal |
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunomicroscopy.Specifications
IgG (H+L) | |
Flow Cytometry 1:20-1:2000, Immunomicroscopy 1:20-1:2000 | |
Polyclonal | |
Goat | |
IgG | |
Affinity purified | |
Based on IEP, this antibody reacts with heavy gamma chains on mouse IgG and light chains on all mouse immunoglobulins. This antibody has been pre-adsorbed against bovine, goat, human, rabbit, and rat IgG. |
4.5mg/mL | |
Flow Cytometry, Immunomicroscopy | |
Unconjugated | |
Adaptive Immunity, Immunology | |
Secondary | |
Reconstitute with sterilized deionized water. Centrifuge to remove any particulates. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title