Learn More
Goat F(ab')2 anti-Mouse IgM Heavy Chain Secondary Antibody, Alkaline Phosphatase, R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgM Heavy Chain |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alkaline Phosphatase (AP) |
| Dilution | Western Blot 1:2000 - 1:10000, ELISA 1:5000, Immunohistochemistry 1:200 - 1:1000 |
| Gene Alias | Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain |
| Gene Symbols | IGHM |
| Host Species | Goat |
| Immunogen | Goat F(ab')2 anti-Mouse IgM Heavy Chain Secondary Antibody was produced by repeated immunization with Mouse IgM heavy chain in goat. |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.