Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat F(ab')2 anti-Mouse IgM Heavy Chain Secondary Antibody, Novus Biologicals™
SDP

Catalog No. NB1205925 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1205925 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1205925 Supplier Novus Biologicals Supplier No. NB1205925
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgM Heavy Chain Polyclonal specifically detects IgM Heavy Chain in Mouse samples. It is validated for ELISA, Immunohistochemistry, Western Blot.

Specifications

Antigen IgM Heavy Chain
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:200 - 1:2000, ELISA 1:2000 - 1:8000, Immunohistochemistry 1:1000 - 1:5000
Gene Alias Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain
Gene Symbols IGHM
Host Species Goat
Immunogen Goat F(ab')2 anti-Mouse IgM Heavy Chain Secondary Antibody was produced by repeated immunization with Mouse IgM heavy chain in goat.
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3507
Test Specificity No reaction was observed against anti-Pepsin or anti-Goat IgG F(c). Specificity was confirmed by ELISA at less than 1% cross-reactivity against other mouse heavy or light chain isotypes.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.