Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat F(ab')2 anti-Rabbit IgG (H+L) Secondary Antibody (Pre-adsorbed), Novus Biologicals™
SDP

Catalog No. NBP260682 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP260682 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP260682 Supplier R&D Systems Supplier No. NBP260682

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Goat Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for ELISA, Immunohistochemistry, SDS-Page, Western Blot.

Specifications

Antigen IgG (H+L)
Applications ELISA, Immunohistochemistry, SDS-Page, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, ELISA 1:20000, Immunohistochemistry 1:1000-1:5000, SDS-Page
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Host Species Goat
Immunogen Rabbit IgG whole molecule
Purification Method Affinity Purified
Quantity 500 μg
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity No reaction was observed against anti-Pepsin, anti-Goat IgG F(c) or Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Mouse, Rat and Sheep Serum Proteins.
Target Species Rabbit
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.