Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat F(ab')2 anti-Rat IgG F(ab')2 Secondary Antibody, PE (Pre-adsorbed), Novus Biologicals™
SDP

Catalog No. NB1206519 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1206519 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1206519 Supplier Novus Biologicals Supplier No. NB1206519
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen IgG F(ab')2
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration LYOPH
Conjugate PE
Dilution Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:250
Host Species Goat
Immunogen Rat IgG F(ab')2 fragment.
Purification Method Affinity Purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity No reaction was observed against anti-Pepsin, anti-Goat IgG F(c), Rat IgG F(c) or Bovine, Horse and Human Serum Proteins.
Target Species Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.