Learn More
Goat F(ab')2 anti-Rat IgG Fc Secondary Antibody, PE (Pre-adsorbed), R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgG Fc |
| Applications | Flow Cytometry, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | PE |
| Dilution | Flow Cytometry 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 - 1:250 |
| Host Species | Goat |
| Immunogen | Goat F(ab')2 anti-Rat IgG Fc fragment Secondary Antibody (Pre-adsorbed) was produced by repeated immunization with Rat F(c) fragment in goat. |
| Purification Method | Affinity Purified |
| Quantity | 0.25 mg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.