Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat IgG HRP-conjugated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. HAF109 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HAF109 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. HAF109 Supplier R&D Systems Supplier No. HAF109
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Donkey Polyclonal Antibody has been used in 36 publications

IgG Polyclonal specifically detects IgG in Goat samples. It is validated for Simple Western∣Western Blot.

Specifications

Antigen IgG
Applications Simple Western, Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, Simple Western
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Donkey
Immunogen Goat IgG
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity Detects goat IgG heavy and light chains in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with human IgG, rabbit IgG, mouse IgG and less than 1% cross-reactivity with chicken IgY is observed.
Target Species Goat
Content And Storage Do not freeze. 6 months from date of receipt, 2 to 8 degreesC as supplied.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.