Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat IgG Isotype Control, HRP, Novus Biologicals™
SDP

Catalog No. NB001856 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001856 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001856 Supplier Novus Biologicals Supplier No. NB41028088H500UL
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

IgG Polyclonal specifically detects IgG in Goat samples. It is validated for Flow Cytometry, ELISA, Fluorophore-linked immunosorbent assay.

Specifications

Antigen IgG
Applications Flow Cytometry, ELISA
Classification Polyclonal
Conjugate HRP
Dilution Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Fluorophore-linked immunosorbent assay 1:100 - 1:2000
Formulation 50 mM Sodium Borate.
Gene Symbols IGHG1
Host Species Goat
Quantity 500 μL
Gene ID (Entrez) 3500
Target Species Goat
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.