Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat IgG VisUCyte HRP Polymer, R&D Systems™

Catalog No. VC004025
Change view
Click to view available options
Quantity:
125 mL
25 mL
250 mL (2 x 125 mL)
50 mL (2 x 25 mL)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
VC004025 25 mL
VC004050 50 mL (2 x 25 mL)
VC004125 125 mL
VC004250 250 mL (2 x 125 mL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. VC004025 Supplier R&D Systems Supplier No. VC004025
This item is not returnable. View return policy

Donkey Polyclonal Antibody has been used in 2 publications

IgG Polyclonal specifically detects IgG in Goat samples. It is validated for Immunohistochemistry.

Specifications

Antigen IgG
Applications Immunohistochemistry
Classification Polyclonal
Conjugate HRP Polymer
Dilution Immunohistochemistry
Formulation Supplied as a solution in PBS with preservative.
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Donkey
Immunogen Goat IgG
Purification Method Affinity Purified
Quantity 25 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity Detects heavy ( gamma ) chains on goat IgG and light chains on all goat immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin goat serum proteins or serum proteins from human, mouse or rat, based on IEP. VisUCyte™ HRP Polymer provides high sensitivity with low background in IHC.
Content And Storage 6 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.